EMAIL THIS PAGE TO A FRIEND
Lung cancer (Amsterdam, Netherlands)
Heterogeneity of circulating tumor cells (CTCs) in patients with recurrent small cell lung cancer (SCLC) treated with pazopanib.
PMID 28212995
Abstract
To investigate the effect of pazopanib on different CTCs subpopulations in patients with recurrent SCLC and evaluate their clinical relevance. Peripheral blood was obtained before the administration of pazopanib (n=56 patients), after the first cycle (n=35 patients) and at disease progression (n=45 patients). CTCs were detected by CellSearch and double immunofluorescent staining using antibodies against the cytokeratins (CK), TTF-1, CD56 and VEGFR2. Before treatment, CTCs could be detected in 50% of patients by CellSearch; phenotypic characterization of CTCs demonstrated that 50%, 46.6% and 27.6% of patients had CD45(-)/TTF1(+), CD45(-)/CD56(+) and TTF-1(+)/CD56(+) CTCs, respectively. Additionally, 59% of CTCs were TTF-1(+)/VEGFR2(+) and 53% CK(+)/VEGFR2(+). One pazopanib cycle resulted to a significant decrease of the number of CTCs (CellSearch: p=0.043) and CK(+)/VEGFR2(+) cells (p=0.027). At the time of PD, both the total number of CTCs (p=0.027) and the number of the different subpopulations were significantly increased compared to post-1st cycle values; this increased CTCs number was associated with a significant increase of TTF-1(+)/VEGFR2(+) (p=0.028) and CK(+)/VEGFR2(+) CTCs (p=0.018). In multivariate analysis, only the number of CTCs as assessed by CellSearch after one treatment cycle was significantly associated with OS (HR: 0.21; p=0.005). Pazopanib has a significant effect on different subpopulations of CTCs in patients with relapsed SCLC; the detection of VEGFR2(+) CTCs during treatment could be a surrogate marker associated with resistance to pazopanib.