EMAIL THIS PAGE TO A FRIEND
FEBS letters
Enhanced CRISPR/Cas9-mediated biallelic genome targeting with dual surrogate reporter-integrated donors.
PMID 28214366
Abstract
The clustered regularly interspaced short palindromic repeat (CRISPR)/CRISPR-associated protein 9 (Cas9) system has recently emerged as a simple, yet powerful genome engineering tool, which has been widely used for genome modification in various organisms and cell types. However, screening biallelic genome-modified cells is often time-consuming and technically challenging. In this study, we incorporated two different surrogate reporter cassettes into paired donor plasmids, which were used as both the surrogate reporters and the knock-in donors. By applying our dual surrogate reporter-integrated donor system, we demonstrate high frequency of CRISPR/Cas9-mediated biallelic genome integration in both human HEK293T and porcine PK15 cells (34.09% and 18.18%, respectively). Our work provides a powerful genetic tool for assisting the selection and enrichment of cells with targeted biallelic genome modification.