EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Diels-Alder reactions of myrcene using intensified continuous-flow reactors.
PMID 28228853
Abstract
This work describes the Diels-Alder reaction of the naturally occurring substituted butadiene, myrcene, with a range of different naturally occurring and synthetic dienophiles. The synthesis of the Diels-Alder adduct from myrcene and acrylic acid, containing surfactant properties, was scaled-up in a plate-type continuous-flow reactor with a volume of 105 mL to a throughput of 2.79 kg of the final product per day. This continuous-flow approach provides a facile alternative scale-up route to conventional batch processing, and it helps to intensify the synthesis protocol by applying higher reaction temperatures and shorter reaction times.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238198
Dimethyl maleate, 96%
C6H8O4
W276200
Myrcene, ≥95%, stabilized, FCC, FG
C10H16