EMAIL THIS PAGE TO A FRIEND
Zhonghua liu xing bing xue za zhi = Zhonghua liuxingbingxue zazhi
[Changes of resistant phenotype and CRISPR/Cas system of four Shigella strains passaged for 90 times without antibiotics].
PMID 28231673
Abstract
Objective: To explore the stability of resistant phenotypes and changes of clustered regularly interspaced short palindromic repeats (CRISPR)/CRISPR-associated (Cas) gene system on four Shigella strains in the absence of antibiotics. Methods: Four clinical isolated Shigella strains that resistant to different antibiotics were consecutive passaged for 90 times without antibiotics. Agar dilution method was used to determine the minimum inhibitory concentration of Shigella strains. After sequence analysis with PCR, CRISPR Finder and Clustal X 2.1 were applied to identify the changes of CRISPR loci in the Shigella strains. Results: After the consecutive transfer of 90 generations, sensitivity to certain antibiotics of four Shigella strains with different drug resistant spectrums increased. Mel-sf1998024/zz resistance to ampicillin, cephalexin, cefotaxime, chloramphenicol decreased, mel-s2014026/sx resistance to norfloxacin, trimethoprim decreased, mel-sf2004004/sx drug resistance to ampicillin, cefuroxime, cefotaxime, chloramphenicol, trimethoprim decreased and mel-sf2013004/bj resistance to chloramphenicol decreased. The spacer of which matched gene codes Cas and its upstream repeat in 3'end of CRISPR3 got lost in mel-sf1998024/zz and mel-sf2013004/bj. Conclusions:Shigella strains could reduce or lose their resistance to some antibiotics after consecutive transfers, without the interference of antibiotics. CRISPR3 locus had dynamic spacers in Shigella strains while CRISPR3 locus and cas genes might have been co-evolved.