EMAIL THIS PAGE TO A FRIEND
Cell
SnapShot: Class 1 CRISPR-Cas Systems.
PMID 28235204
Abstract
Class 1 CRISPR-Cas systems are characterized by effector modules consisting of multiple subunits. Class 1 systems comprise about 90% of all CRISPR-Cas loci identified in bacteria and archaea and can target both DNA and RNA.