EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Synthesis of novel monomeric graphene quantum dots and corresponding nanocomposite with molecularly imprinted polymer for electrochemical detection of an anticancerous ifosfamide drug.
PMID 28237900
Abstract
This paper reports a typical synthesis of a nanocomposite of functionalized graphene quantum dots and imprinted polymer at the surface of screen-printed carbon electrode using N-acryloyl-4-aminobenzamide, as a functional monomer, and an anticancerous drug, ifosfamide, as a print molecule (test analyte). Herein, graphene quantum dots in nanocomposite practically induced the electrocatalytic activity by lowering the oxidation overpotential of test analyte and thereby amplifying electronic transmission, without any interfacial barrier in between the film and the electrode surface. The differential pulse anodic stripping signal at functionalized graphene quantum dots based imprinted sensor was realized to be about 3- and 7-fold higher as compared to the traditionally made imprinted polymers prepared in the presence and the absence of graphene quantum dots (un-functionalized), respectively. This may be attributed to a pertinent synergism in between the positively charged functionalized graphene quantum dots in the film and the target analyte toward the enhancement of electro-conductivity of the film and thereby the electrode kinetics. In fact, the covalent attachment of graphene quantum dots with N-acryloyl-4-aminobenzamide molecules might exert an extended conjugation at their interface facilitating electro conducting to render the channelized pathways for the electron transport. The proposed sensor is practically applicable to the ultratrace evaluation of ifosfamide in real (biological/pharmaceutical) samples with detection limit as low as 0.11ngmL(-1) (S/N=3), without any matrix effect, cross-reactivity, and false-positives.