EMAIL THIS PAGE TO A FRIEND
Basic research in cardiology
Leukocyte iNOS is required for inflammation and pathological remodeling in ischemic heart failure.
PMID 28238121
Abstract
In the failing heart, iNOS is expressed by both macrophages and cardiomyocytes. We hypothesized that inflammatory cell-localized iNOS exacerbates left ventricular (LV) remodeling. Wild-type (WT) C57BL/6 mice underwent total body irradiation and reconstitution with bone marrow from iNOS(-/-) mice (iNOS(-/-)c) or WT mice (WTc). Chimeric mice underwent coronary ligation to induce large infarction and ischemic heart failure (HF), or sham surgery. After 28xa0days, as compared with WTc sham mice, WTc HF mice exhibited significant (pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2551 Brewer thioglycollate medium, for microbiology