PloS one
Prognostic impact of pretherapeutic gamma-glutamyltransferase on patients with nasopharyngeal carcinoma.
PMID 28241022
Abstract
Gamma-glutamyltransferase (GGT) is a membrane-bound enzyme involved in the metabolism of glutathione. Studies suggested that GGT played an important role in the tumor development, progression, invasion and drug resistance and prognosis. The association between GGT and prognosis of patients with nasopharyngeal carcinoma (NPC) was unknown. This study was conducted to investigate the association of pretherapeutic serum level of GGT with clinical-pathological parameters and survival in patients with NPC. Two hundred and twenty-two patients with NPC were recruited in this study and were stratified into two GGT risk groups (≤ 34.5 U/L, > 34.5 U/L). The association of pretherapeutic serum GGT levels with clinical-pathological parameters was examined. Univariate and multivariate survival analyses were performed. The pretherapeutic serum level of GGT was not associated with gender, age, pathology, T stage, N stage, TNM stage, chemotherapy or radiotherapy in patients with NPC. Patients in the high-risk GGT group had a poorer survival than the low-risk GGT group (3-year overall survival, 74.2% vs. 50.2%, P = 0.001; 3-year progression-free survival, 76.4% vs. 47.1%, P < 0.001; 3-year loco-regional relapse-free survival, 76.4% vs. 51.3%, P < 0.001; 3-year distant metastasis-free survival, 89.5% vs. 66.4%, P < 0.001). Multivariate analysis suggested that patients in the high-risk GGT group had 2.117 (95% confidence interval [CI], 1.225 ∼ 3.659, P = 0.007) times the risk of death, 2.836 (95% CI, 1.765 ∼ 4.557, P < 0.001) times the risk of progression, 2.551 (95% CI, 1.573 ∼ 4.138, P < 0.001) times the risk of relapse, and 3.331 (95% CI, 1.676 ∼ 6.622, P < 0.001) times the risk of metastasis compared with those in the low-risk GGT group. The pretherapeutic serum level of GGT might serve as a novel independent prognostic factor for overall-survival, progression-free survival, loco-regional relapse-free survival and distant metastasis-free survival in patients with NPC.