EMAIL THIS PAGE TO A FRIEND
World journal of microbiology & biotechnology
Bioactive compounds of Aspergillus terreus-F7, an endophytic fungus from Hyptis suaveolens (L.) Poit.
PMID 28243983
Abstract
The compounds terrein (1), butyrolactone I (2), and butyrolactone V (3) were isolated from the ethyl acetate extract (EtOAc) of the endophytic fungus Aspergillus terreus-F7 obtained from Hyptis suaveolens (L.) Poit. The extract and the compounds presented schistosomicidal activity against Schistosoma mansoni; at 100xa0µg/mL for EtOAc extract, 1297.3xa0µM for compound 1, 235.6xa0µM for compound 2, and 454.1xa0µM for compound 3, they killed 100% of the parasites after 72xa0h of treatment. Compounds 1, 2, and 3 exerted moderate leishmanicidal activity against Leishmania amazonensis (IC50 ranged from 23.7 to 78.6xa0µM). At 235.6 and 227.0xa0µM, compounds 2 and 3, respectively, scavenged 95.92 and 95.12% of the DPPH radical (2,2-diphenyl-1-picryl-hydrazyl), respectively. Regarding the cytotoxicity against the breast tumor cell lines MDA-MB-231 and MCF-7, compound 2 gave IC50 of 34.4 and 17.4xa0µM, respectively, while compound 3 afforded IC50 of 22.2 and 31.9xa0µM, respectively. At 117.6xa0µM, compound 2 inhibited the growth of and killed the pathogen Escherichia coli (ATCC 25922). Compounds 1, 2, and 3 displayed low toxicity against the normal line of human lung fibroblasts (GM07492A cells), with IC50 of 15.3 × 10(3), 3.4 × 10(3), and 5.8 × 10(3)xa0µM, respectively. This is the first report on (i) the in vitro schistosomicidal and leishmanicidal activities of the EtOAc extract of A. terreus-F7 and compounds 1, 2, and 3; and (ii) the antitumor activity of compounds 2 and 3 against MDA-MB-231 and MCF-7 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774243
1,3-Propylene sulfite, 99%
C3H6O3S