EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
Precise base editing in rice, wheat and maize with a Cas9-cytidine deaminase fusion.
PMID 28244994
Abstract
Targeted base editing in plants without the need for a foreign DNA donor or double-stranded DNA cleavage would accelerate genome modification and breeding in a wide array of crops. We used a CRISPR-Cas9 nickase-cytidine deaminase fusion to achieve targeted conversion of cytosine to thymine from position 3 to 9 within the protospacer in both protoplasts and regenerated rice, wheat and maize plants at frequencies of up to 43.48%.