EMAIL THIS PAGE TO A FRIEND
Genome biology
Knocking out consumer concerns and regulator's rules: efficient use of CRISPR/Cas ribonucleoprotein complexes for genome editing in cereals.
PMID 28245842
Abstract
Selection-free genome editing using Cas9 ribonucleoprotein embryo bombardment has been achieved for maize and wheat. This is a breakthrough that should make new breeding technologies more acceptable for worldwide use.