EMAIL THIS PAGE TO A FRIEND
Journal of biomedical optics
Noninvasive fluence rate mapping in living tissues using magnetic resonance thermometry.
PMID 28246674
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2500 Gelatin from porcine skin, gel strength 300, Type A