EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
The role of cytochrome P450 1B1 and its associated mid-chain hydroxyeicosatetraenoic acid metabolites in the development of cardiac hypertrophy induced by isoproterenol.
PMID 28251434
Abstract
Numerous experimental studies have demonstrated the role of cytochrome P450 1B1 (CYP1B1) and its associated mid-chain hydroxyeicosatetraenoic acids (mid-chain HETEs) metabolite in the pathogenesis of cardiac hypertrophy. However, the ability of isoproterenol (ISO) to induce cardiac hypertrophy through mid-chain HETEs has not been investigated yet. Therefore, we hypothesized that ISO induces cardiac hypertrophy through the induction of CYP1B1 and its associated mid-chain HETE metabolites. To test our hypothesis, Sprague-Dawley rats were treated with ISO (5xa0mg/kg i.p.) for 12 and 72xa0h whereas, human ventricular cardiomyocytes RL-14 cells were exposed to 100xa0μM ISO in the presence and absence of 0.5xa0μM tetramethoxystilbene (TMS) a selective CYP1B1 inhibitor, or 25 nM CYP1B1-siRNA. Moreover, RL-14 cells were transiently transfected with the CRISPR-CYP1B1 plasmid. Thereafter, real-time PCR, western blot analysis, and liquid chromatography-electrospray ionization mass spectroscopy were used to determine the level of gene expression, protein expression, and mid-chain HETEs, respectively. Our results showed that ISO induced CYP1B1 protein expression and the level of cardiac mid-chain HETEs in vivo at pre-hypertrophic and hypertrophic stage. In vitro, inhibition of CYP1B1 using TMS or CYP1B1-siRNA significantly attenuates ISO-induced hypertrophy. Furthermore, overexpression of CYP1B1 significantly induced cellular hypertrophy and mid-chain HETEs metabolite. Mechanistically, the protective effect of TMS against cardiac hypertrophy was mediated through the modulation of superoxide anion, mitogen-activated protein kinases (MAPKs), and nuclear factor-κB (NF-κB). In conclusion, our study provides the first evidence that CYP1B1 and its associated mid-chain HETE metabolites are directly involved in the ISO-induced cardiac hypertrophy.