EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Re-evaluation of thyroid hormone signaling antagonism of tetrabromobisphenol A for validating the T3-induced Xenopus metamorphosis assay.
PMID 28254054
Abstract
We developed the T3-induced Xenopus metamorphosis assay, which is supposed to be able to sensitively detect thyroid hormone (TH) signaling disruption of chemicals. The present study aimed to validate the T3-induced Xenopus metamorphosis assay by re-evaluating the TH signaling antagonism of tetrabromobisphenol A (TBBPA), a known TH signaling disruptor. According to the assay we developed, Xenopus tadpoles at stage 52 were exposed to 10-500nmol/L TBBPA in the presence of 1nmol/L T3. After 96hr of exposure, TBBPA in the range of 10-500nmol/L was found to significantly inhibit T3-induced morphological changes of Xenopus tadpoles in a concentration-dependent manner in term of body weight and four morphological endpoints including head area (HA), mouth width (MW), unilateral brain width/brain length (ULBW/BL), and hind-limb length/snout-vent length (HLL/SVL). The results show that these endpoints we developed are sensitive for characterizing the antagonistic effects of TBBPA on T3-induced metamorphosis. Following a 24-hr exposure, we found that TBBPA antagonized expression of T3-induced TH-response genes in the tail, which is consistent with previous findings in the intestine. We propose that the tail can be used as an alternative tissue to the intestine for examining molecular endpoints for evaluating TH signaling disruption. In conclusion, our results demonstrate that the T3-induced Xenopus metamorphosis assay we developed is an ideal in vivo assay for detecting TH signaling disruption.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E10505
Ethyl 3-aminobenzoate, 97%
C9H11NO2