EMAIL THIS PAGE TO A FRIEND
BMC evolutionary biology
Comparative analyses of glycerotoxin expression unveil a novel structural organization of the bloodworm venom system.
PMID 28259138
Abstract
We present the first molecular characterization of glycerotoxin (GLTx), a potent neurotoxin found in the venom of the bloodworm Glycera tridactyla (Glyceridae, Annelida). Within the animal kingdom, GLTx shows a unique mode of action as it can specifically up-regulate the activity of Cav2.2 channels (N-type) in a reversible manner. The lack of sequence information has so far hampered a detailed understanding of its mode of action. Our analyses reveal three ~3.8xa0kb GLTx full-length transcripts, show that GLTx represents a multigene family, and suggest it functions as a dimer. An integrative approach using transcriptomics, quantitative real-time PCR, in situ hybridization, and immunocytochemistry shows that GLTx is highly expressed exclusively in four pharyngeal lobes, a previously unrecognized part of the venom apparatus. Our results overturn a century old textbook view on the glycerid venom system, suggesting that it is anatomically and functionally much more complex than previously thought. The herein presented GLTx sequence information constitutes an important step towards the establishment of GLTx as a versatile tool to understand the mechanism of synaptic function, as well as the mode of action of this novel neurotoxin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469548
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
C8H11F3O2