EMAIL THIS PAGE TO A FRIEND
JACC. Cardiovascular interventions
Bioresorbable Everolimus-Eluting Vascular Scaffold for Long Coronary Lesions: A Subanalysis of the International, Multicenter GHOST-EUxa0Registry.
PMID 28259663
Abstract
The authors sought to investigate 1-year outcomes in patients treated with bioresorbable everolimus-elutingxa0vascular scaffolds (BVS) for "long coronary lesions." The present substudy derived from the GHOST-EU registry included 1,722 lesions in 1,468 consecutive patients, enrolled between November 2011 and September 2014 at 11 European centers. The lesions were divided into 3 groups according to continuous BVS length: 1) shorter than 30 mm; 2) between 30 and 60 mm; and 3) longer than 60 mm. Primary device-oriented endpoint (target lesion failure [TLF]) was defined as a combination of cardiovascular death, target vessel myocardial infarction, or clinically driven target lesion revascularization. Patients with lesionsxa0≥60 mm had more comorbidities and more complex lesion characteristics, including chronic total occlusions (37%), bifurcation lesions (40.3%), higher Syntax score (16.4 ± 7.8), and higher number of scaffolds implanted per lesion (3.3 ± 0.9 mm). The main target vessel was the left anterior coronary artery in all groups. Median follow-up was 384 (interquartile range: 359 to 459) days. One-year follow-up was completed in 70.3% of patients. TLF at 1 year was significantly higher in group C (group A 4.8%, group B 4.5%, group C 14.3%; overall pxa0=xa00.001), whereas there were no significant differences between groups A and B. Finally, a numerically higher (but not statistically significant) number of scaffold thromboses were observed in group C when compared with shorter lesions (group A 2.1%, group B 1.1%, group C 3.8%; overall pxa0= 0.29). In a real-world setting, treatment of long coronary lesions with BVSxa0≥60 mm was associated withxa0axa0higher TLF rate, driven by myocardial infarction and clinically driven target lesion revascularization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0331
Prasugrel, ≥98% (HPLC)
C20H20FNO3S