EMAIL THIS PAGE TO A FRIEND
Nature biotechnology
GuideScan software for improved single and paired CRISPR guide RNA design.
PMID 28263296
Abstract
We present GuideScan software for the design of CRISPR guide RNA libraries that can be used to edit coding and noncoding genomic regions. GuideScan produces high-density sets of guide RNAs (gRNAs) for single- and paired-gRNA genome-wide screens. We also show that the trie data structure of GuideScan enables the design of gRNAs that are more specific than those designed by existing tools.