EMAIL THIS PAGE TO A FRIEND
Natural product research
Fragrance components of Platanthera bifolia subsp. osca.
PMID 28278677
Abstract
SPME-GC-MS analysis of the scent of Platanthera bifolia subsp. osca collected during the night showed as main components lilac alcohols B, C and D and lilac aldehydes A, B and C. Other significant chemical components were linalool and caryophyllene. Some differences were found in comparison with previously reported analyses of the scent of P. bifolia and Platanthera chlorantha. The most important difference found was in the composition of the ester fraction.