EMAIL THIS PAGE TO A FRIEND
International journal of immunopathology and pharmacology
Protective effect of medicinal fungus Xylaria nigripes mycelia extracts against hydrogen peroxide-induced apoptosis in PC12 cells.
PMID 28281874
Abstract
Xylaria nigripes ( XN) is a medicinal fungus that was used traditionally as a diuretic, nerve tonic, and for treating insomnia and trauma. In this study, we elucidated possible mechanisms of neuroprotective effects of XN mycelia extracts. XN mycelia were produced by fermentation. Hot water extract and 70% ethanol extract of XN mycelia were evaluated on hydrogen peroxide (H2O2)-induced apoptosis in PC12, a rat pheochromocytoma cell line. Both XN extracts effectively protected PC12 cells against H2O2-induced cell damage by inhibiting release of lactate dehydrogenase, decreasing DNA damage, restoring mitochondrial membrane potential, and arresting abnormal apoptosis through upregulation of Bcl-2 and downregulation of Bax and caspase 3. Compared to water extract, ethanol extract showed not only greater neuroprotective effects but also a higher antioxidant activity by scavenging DPPH radicals, inhibiting lipid peroxidation, and reducing power. High phenolic content and antioxidant activity may provide the neuroprotective properties of XN ethanol extract.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
673099
Biphenyl-4,4′-dithiol, 95%
C12H10S2