EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Lower concentrations of receptor for advanced glycation end products and epiregulin in amniotic fluid correlate to chemically induced cleft palate in mice.
PMID 28282589
Abstract
This study investigated the correlation between differentially expressed proteins in amniotic fluid (AF) and cleft palate induced by all-trans retinoic acid (atRA), and 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) in mice. Seven proteins were differentially expressed at embryonic day (E) 16.5 in atRA and control groups as revealed by label-based mouse antibody array. Enzyme-linked immunosorbent assay was further used to detect the expression levels of these proteins in AF from E13.5 to E16.5 in atRA, TCDD, and control groups. The cleft palate groups showed lower concentrations of receptor for advanced glycation end products (RAGE) and epiregulin at E16.5. RAGE immunostaining obviously decreased in palatal tissue sections obtained from E14.5 to E16.5 in the cleft palate groups as revealed by immunohistochemistry. These findings indicate that reduced levels of RAGE and epiregulin in AF are correlated to chemically induced cleft palate in mice.
Related Materials