EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Hot-Water Extracts from Roots of Vitis thunbergii var. taiwaniana and Identified ε-Viniferin Improve Obesity in High-Fat Diet-Induced Mice.
PMID 28285527
Abstract
In this study, hot-water extracts (HW) from roots of Vitis thunbergii var. taiwaniana (VTT-R) were shown to lower levels of lipid accumulation significantly (P < 0.01 or 0.001) compared to the control in 3T3-L1 adipocytes. The VTT-R-HW (40 mg/kg) interventions concurrent with a high-fat (HF) diet in C57BL/6 mice over a 5 eek period were shown to reduce body weights significantly (P < 0.05) compared to those of mice fed a HF diet under the same food-intake regimen. The (+)-ε-viniferin isolated from VTT-R-HW was shown to reduce the size of lipid deposits significantly compared to the control (P < 0.05 or 0.001) in 3T3-L1 adipocytes, and dose-dependent 3-hydroxy-3-methylglutaryl-CoA (HMG-CoA) reductase inhibitions showed that the 50% inhibitory concentration was calculated to be 96 μM. The two-stage (+)-ε-viniferin interventions (10 mg/kg, day 1 to day 38; 25 mg/kg, day 39 to day 58) were shown to lower mice body weights significantly (P < 0.05 or 0.001), the weight ratio of mesenteric fat, blood glucose, total cholesterol, and low-density lipoprotein compared to that of the HF group under the same food-intake regimen but without concurrent VTT-R-HW interventions. It might be possible to use VTT-R-HW or (+)-ε-viniferin as an ingredient in the development of functional foods for weight management, and this will need to be investigated further.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H6132
DL-3-Hydroxy-3-methylglutaryl coenzyme A sodium salt hydrate, ≥90% (HPLC)
C27H42N7Na2O20P3S · xH2O