EMAIL THIS PAGE TO A FRIEND
Food research international (Ottawa, Ont.)
Peptides from gluten digestion: A comparison between old and modern wheat varieties.
PMID 28290332
Abstract
Coeliac disease is an autoimmune enteropathy that develops in genetically predisposed subjects after the ingestion of gluten or related proteins. Coeliac disease has an increasing incidence in the last years in western countries and it has been suggested that wheat breeding might have contributed to select more toxic forms of gluten. In this work, we analysed gluten peptides generated by in vitro digestion of different old and modern Triticum varieties, using LC-MS. We concluded that old varieties analysed produced a higher quantity of peptides containing immunogenic and toxic sequences than modern ones. Thus old wheat lines are not to be considered "safer" for subjects that are genetically predisposed to celiac disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47636
Fmoc-Pro-OH, ≥99.0% (HPLC)
C20H19NO4
12804
HBTU, ≥98.0% (T)
C11H16F6N5OP