EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Activin-A co-opts IRF4 and AhR signaling to induce human regulatory T cells that restrain asthmatic responses.
PMID 28320933
Abstract
Type 1 regulatory T (Tr1) cells play a pivotal role in restraining human T-cell responses toward environmental allergens and protecting against allergic diseases. Still, the precise molecular cues that underlie their transcriptional and functional specification remain elusive. Here, we show that the cytokine activin-A instructs the generation of CD4(+) T cells that express the Tr1-cell-associated molecules IL-10, inducible T-Cell costimulator (ICOS), lymphocyte activation gene 3 protein (LAG-3), and CD49b, and exert strongly suppressive functions toward allergic responses induced by naive and in vivo-primed human T helper 2 cells. Moreover, mechanistic studies reveal that activin-A signaling induces the activation of the transcription factor interferon regulatory factor (IRF4), which, along with the environmental sensor aryl hydrocarbon receptor, forms a multipartite transcriptional complex that binds in IL-10 and ICOS promoter elements and controls gene expression in human CD4(+) T cells. In fact, IRF4 silencing abrogates activin-A-driven IL10 and ICOS up-regulation and impairs the suppressive functions of human activin-A-induced Tr1-like (act-A-iTr1) cells. Importantly, using a humanized mouse model of allergic asthma, we demonstrate that adoptive transfer of human act-A-iTr1 cells, both in preventive and therapeutic protocols, confers significant protection against cardinal asthma manifestations, including pulmonary inflammation. Overall, our findings uncover an activin-A-induced IRF4-aryl hydrocarbon receptor (AhR)-dependent transcriptional network, which generates suppressive human Tr1 cells that may be harnessed for the control of allergic diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0447
SIS3, ≥98% (HPLC), powder
C28H27N3O3 · HCl