EMAIL THIS PAGE TO A FRIEND
Scientific reports
12-O-Tetradecanoylphorbol-13-acetate (TPA) is anti-tumorigenic in liver cancer cells via inhibiting YAP through AMOT.
PMID 28322318
Abstract
TPA stimulates carcinogenesis in various types of cancers. However, we found that TPA inhibits transformative phenotypes in liver cancer cells via the translocation of YAP from the nucleus, where it functions as a transcriptional co-factor, to the cytoplasm. Such effects led to a separation of YAP from its dependent transcription factors. The inhibitory effects of TPA on YAP were AMOT dependent. Without AMOT, TPA was unable to alter YAP activity. Importantly, the depletion of YAP and AMOT blocked the TPA-reduced transformative phenotypes. In sum, TPA has been established as an anti-tumorigenic drug in liver cancer cells via YAP and AMOT.