EMAIL THIS PAGE TO A FRIEND
Biotechnology advances
New strategies and approaches for engineering biosynthetic gene clusters of microbial natural products.
PMID 28323062
Abstract
With the rapidly growing number of sequenced microbial (meta)genomes, enormous cryptic natural product (NP) biosynthetic gene clusters (BGCs) have been identified, which are regarded as a rich reservoir for novel drug discovery. A series of powerful tools for engineering BGCs has accelerated the discovery and development of pharmaceutically active NPs. Here, we describe recent advances in the strategies for BGCs manipulation, which are driven by emerging technologies, including efficient DNA recombination systems, versatile CRISPR/Cas9 genome editing tools and diverse DNA assembly methods. We further discuss how these approaches could be used for genome mining studies and industrial strain improvement.