EMAIL THIS PAGE TO A FRIEND
Insect biochemistry and molecular biology
Afidopyropen: New and potent modulator of insect transient receptor potential channels.
PMID 28347703
Abstract
The commercial insecticides pymetrozine and pyrifluquinazon control plant-sucking pests by disturbing their coordination and ability to feed. We have previously shown that these compounds act by overstimulating and eventually silencing vanilloid-type transient receptor potential (TRPV) channels, which consist of two proteins, Nanchung and Inactive, that are co-expressed exclusively in insect chordotonal stretch receptor neurons. Here we show that a new insecticidal compound, afidopyropen, modulates chordotonal organs of American grasshoppers (Schistocerca americana) in the same fashion. Afidopyropen stimulated heterologously expressed TRPV channels from two different insect species - fruit fly (Drosophila melanogaster) and pea aphid (Acyrthosiphon pisum) - but did not affect function of the mammalian TRPV channel TRPV4. Activation of the insect TRPVs required simultaneous expression of both Nanchung and Inactive proteins. Tritium-labeled afidopyropen bound fruit fly TRPVs with higher affinity than pymetrozine and competed with pymetrozine for binding. Nanchung protein formed the main binding interface for afidopyropen, whereas co-expression of Inactive dramatically increased binding affinity. Another modulator of chordotonal organs, flonicamid, did not activate insect TRPV channels, nor did it compete with afidopyropen for binding, indicating that it has a different target site. These results define afidopyropen as a new, potent and specific modulator of insect TRPV channels, and provide insight into the unique binding mode of these compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46119
Pymetrozin, PESTANAL®, analytical standard
C10H11N5O