EMAIL THIS PAGE TO A FRIEND
Microbial cell factories
Erratum to: SWITCH: a dynamic CRISPR tool for genome engineering and metabolic pathway control for cell factory construction in Saccharomyces cerevisiae.
PMID 28351356