EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
2D-NMR investigation and in vitro evaluation of antioxidant, antigenotoxic and estrogenic/antiestrogenic activities of strawberry grape.
PMID 28351770
Abstract
Strawberry grape is considered beneficial due to its extensive phytochemical properties. To expand the knowledge about the chemical constituents and the biological activities of the whole plant, 2D-NMR investigation has been carried out on pulp, peel, seeds, stalks and leaves. Catechin and epicatechin were identified as the main constituents of the seed extract, quercetin and ferulic acid were detected in the leaves and malvidin and cyanidin glucopyranoside in the peels. The leaf, stalk and seed extracts were found to be very rich in phytochemicals and were tested for their ability to reduce the mutagenicity and genotoxicity of standard agents via Salmonella mutagenicity assay and SOS chromotest, respectively. Moreover, the estrogen/antiestrogen-like activity was evaluated on the MCF-7 estrogen-responsive cells. Seed and stalk extracts had an elevated antimutagenic/antigenotoxic activity. Stalk extracts highly reduced the proliferative effect of natural estrogen, 17β-estradiol.
Related Materials