EMAIL THIS PAGE TO A FRIEND
Oncology letters
Effect of targeted silencing of IL-8 on in vitro migration and invasion of SKOV3 ovarian cancer cells.
PMID 28356930
Abstract
The aim of the study was to determine whether interleukin-8 (IL-8) affects human SKOV3 ovarian cancer cell migration and invasion by targeting silencing of IL-8 expression. Silencing small-interfering RNA (siRNA) targeting IL-8 gene was constructed to infect SKOV3 cells by lentiviral vector. The expression of IL-8 and p-nuclear factor (NF)-κB protein was detected by western blot analysis. The wound scratch and Transwell tests were used to assay the cell migration and invasiveness of SKOV3 cells infected with lentiviral vector targeting IL-8 gene siRNA. The levels of IL-8 protein expressed by SKOV3 cells infected by lentiviral vector targeting IL-8 gene siRNA decreased by 72.3%. IL-8 (50 ng/ml) increased the ability of SKOV3 cells to suppress cell migration (p<0.01). Cisplatin and silencing of IL-8 achieved the ability to inhibit SKOV3 cell invasion (p<0.01), and 100 ng/ml concentration of IL-8 enhanced the ability of SKOV3 invasion (p<0.01). Silencing of IL-8 to a certain extent reduced the expression of p-NF-κB proteins, but it was not statistically significant. In conclusion, silencing of IL-8 may inhibit the migration and invasion of SKOV3 cells, which may be independent of the p-NF-κB protein.