EMAIL THIS PAGE TO A FRIEND
Pathogens and disease
The CRISPR-Cas9 system in Neisseria spp.
PMID 28369433
Abstract
Bacteria and archaea possess numerous defense systems to combat viral infections and other mobile genetic elements. Uniquely among these, CRISPR-Cas (clustered, regularly interspaced short palindromic repeats-CRISPR associated) provides adaptive genetic interference against foreign nucleic acids. Here we review recent advances on the CRISPR-Cas9 system in Neisseria spp, with a focus on its biological functions in genetic transfer, its mechanistic features that establish new paradigms and its technological applications in eukaryotic genome engineering.