EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Porous CoFe2O4 nanocubes derived from metal-organic frameworks as high-performance anode for sodium ion batteries.
PMID 28371673
Abstract
Recently sodium ion batteries (SIBs) as a new energy storage system have attracted enormous interests. Unfortunately, the development of high-performance electrode materials for SIBs is restricted owing to the large volume change during sodium insertion and extraction. In this work, porous CoFe2O4 nanocubes (PCFO-NCs) were prepared simply by annealing metal-organic frameworks and used as anode materials for SIBs. The PCFO-NCs exhibit a high initial Coulombic efficiency of 68.8% and a maximum reversible capacity of 360mAhg(-1) after 50 cycles at the current density of 50mAg(-1), as well as good rate capability and excellent cycling stability at high current density. The excellent electrochemical performance can be attributed the short diffusion distance of sodium ion due to the good interfacial contact between electrode and electrolyte, and the buffering of volume change during charge/discharge processes by the porous structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757349
Fluoroethylene carbonate, 99%
C3H3FO3