EMAIL THIS PAGE TO A FRIEND
eLife
How should the applications of genome editing be assessed and regulated?
PMID 28375079
Abstract
An EASAC working group on genome editing recommends that regulators should focus on specific applications of these new techniques rather than attempting to regulate genome editing itself as a new technology.