EMAIL THIS PAGE TO A FRIEND
Acta chimica Slovenica
Synthesis, Structures and Antimicrobial Activities of Two Cobalt(II) Complexes [Co(L1)2(OH2)2] and [Co(L2)2].
PMID 28380227
Abstract
A new cobalt(II) complex, [Co(L1)2(OH2)2] (1), was prepared by the reaction of 3-bromo-5-chlorosalicylaldehyde (HL1) with cobalt nitrate in methanol. Reaction of 1 with cyclopropylamine in methanol afforded the Schiff base cobalt(II) complex, [Co(L2)2] (2), where L2 is the deprotonated form of 2-bromo-4-chloro-6-(cyclopropyliminomethyl)phenol (HL2). The complexes have been characterized by elemental analyses, IR spectroscopy, and single-crystal X-ray diffraction. The L1 ligand coordinates to the Co atom through the phenolate O and carbonyl O atoms, while the L2 ligand coordinates to the Co atom through the phenolate O and imino N atoms. The Co atom in complex 1 adopts octahedral coordination and that in complex 2 adopts tetrahedral coordination. The effect of the free ligands and the cobalt complexes on the antimicrobial activities against Staphylococcus aureus, Escherichia coli, and Candida albicans was studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
497592
3-Bromo-5-chlorosalicylaldehyde, 98%
C7H4BrClO2
125504
Cyclopropylamine, 98%
C3H7N