EMAIL THIS PAGE TO A FRIEND
Microbial cell factories
Engineering Halomonas species TD01 for enhanced polyhydroxyalkanoates synthesis via CRISPRi.
PMID 28381263
Abstract
Clustered regularly interspaced short palindromic repeats interference (CRISPRi) has provided an efficient approach for targeted gene inhibition. A non-model microorganism Halomonas species TD01 has been developed as a promising industrial producer of polyhydroxyalkanoates (PHA), a family of biodegradable polyesters accumulated by bacteria as a carbon and energy reserve compound. A controllable gene repression system, such as CRISPRi, is needed for Halomonas sp. TD01 to regulate its gene expression levels. For the first time CRISPRi was successfully used in Halomonas sp. TD01 to repress expression of ftsZ gene encoding bacterial fission ring formation protein, leading to an elongated cell morphology with typical filamentous shape similar to phenomenon observed with Escherichia coli. CRISPRi was employed to regulate expressions of prpC gene encoding 2-methylcitrate synthase for regulating 3-hydroxyvalerate monomer ratio in PHBV copolymers of 3-hydroxybutyrate (HB) and 3-hydroxyvalerate (HV). Percentages of HV in PHBV copolymers were controllable ranging from less than 1 to 13%. Furthermore, repressions on gltA gene encoding citrate synthase channeled more acetyl-CoA from the tricarboxylic acid (TCA) cycle to poly(3-hydroxybutyrate) (PHB) synthesis. The PHB accumulation by Halomonas sp. TD01 with its gltA gene repressed in various intensities via CRISPRi was increased by approximately 8% compared with the wild type control containing the CRISPRi vector without target. It has now been confirmed that the CRISPRi system can be applied to Halomonas sp. TD01, a promising industrial strain for production of various PHA and chemicals under open and continuous fermentation process conditions. In details, the CRISPRi system was successfully designed in this study to target genes of ftsZ, prpC and gltA, achieving longer cell sizes, channeling more substrates to PHBV and PHB synthesis, respectively. CRISPRi can be expected to use for more metabolic engineering applications in non-model organisms.