EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
A Novel Methodology for Synthesis of 1,5,6-Trisubstituted 2(1H)-Pyrazinones of Biological Interest.
PMID 28381677
Abstract
In this report, we describe a new method for the synthesis of densely functionalized 2(1H)-pyrazinones. Treatment of mesoionic 1,3-oxazolium-5-olates with carbanions derived from activated methylene isocyanides (p-toluenesulfonylmethyl isocyanide (TosMIC) and ethyl isocyanoacetate) causes a novel ring transformation affording 2(1H)-pyrazinones in moderate to high yields. The cytotoxicity and antibacterial activity of some of the obtained products were studied and some of the products exhibited tumor-specific cytotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2011
Melphalan, powder
C13H18Cl2N2O2