EMAIL THIS PAGE TO A FRIEND
The FEBS journal
AIDing-targeted protein degradation in Drosophila.
PMID 28382696
Abstract
Conditional protein depletion is highly desirable for investigating protein functions in complex organisms. In this issue, Bence and colleagues combined auxin-inducible degradation with CRISPR, establishing an elegant tool to control protein levels. They achieve precise spatio-temporal control of protein degradation during Drosophila oogenesis and early embryogenesis by combining suitable GAL4 drivers (spatial control) with auxin feeding protocols (temporal control).