EMAIL THIS PAGE TO A FRIEND
The American journal of Chinese medicine
Paris polyphylla Suppresses Proliferation and Vasculogenic Mimicry of Human Osteosarcoma Cells and Inhibits Tumor Growth In Vivo.
PMID 28385078
Abstract
Paris polyphylla, a traditional antipyretic-detoxicate chinese medicinal herb, has been applied extensively in cancer treatments for nearly 2000 years. The purpose of the present study is to evaluate the potential anti-osteosarcoma effects of Paris polyphylla ethanol extract (PPEE) and to investigate its underlying mechanisms. The antiproliferation activity of PPEE was tested on 143B, MG-63, U-2 OS and hFOB1.19 cells using MTT assay. The pro-apoptotic and cell cycle arrest effects of PPEE were confirmed by Hoechst 33342 staining and flow cytometry. The antimigratory, anti-invasive and antivasculogenic mimicry (VM) effects of PPEE were investigated by wound healing, Transwell and 3D culture assays. Mouse xenograft model was used to examine its anti-osteosarcoma efficacy in vivo. Hematologic profiles and hepatorenal functions were evaluated to assess the toxicity of PPEE. PPEE evidently suppressed cell proliferation of 143B, MG-63 and U-2 OS with IC50 values of 10-60[Formula: see text][Formula: see text]g/mL, but showed little cytotoxicity against normal osteoblastic cell. PPEE promoted apoptosis in 143B cell via caspase activation, increased Bax/Bcl-2 ratio and PARP cleavage. It also induced G2/M phase arrest associated with elevated phosphorylation of CDK1, Cdc25C, Chk2 and down-regulation of cyclin B1, CDK1, Cdc25C expression. Additionally, PPEE inhibited 143B cell migration, invasion and VM formation at noncytotoxic concentrations through decreasing the expression of FAK, Mig-7, MMP2 and MMP9. Finally, daily oral administration of PPEE for four weeks exhibits potent antitumor and anti-VM activity in 143B xenograft model with low toxicity. Taken together, these findings demonstrated PPEE possesses anti-osteosarcoma and anti-VM activity in vitro and in vivo, and therefore is a potential candidate for osteosarcoma treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4301213
Anti-MIG7 antibody produced in rabbit, affinity isolated antibody