EMAIL THIS PAGE TO A FRIEND
Nature communications
Papilloma-pseudovirus eradicates intestinal tumours and triples the lifespan of Apc(Min/+) mice.
PMID 28397782
Abstract
Inducing tumour-specific adaptive immunity, such as cytotoxic T lymphocyte (CTL) response, can result in promising antitumour effect against several human malignancies, especially in combination with immune checkpoint blockade strategies. However, little is known whether activation of innate immunity can lead to direct tumoricidal effect. Here, we develop a papilloma pseudovirus-based oral immunotherapeutic approach that shows strong tumoricidal effects in the gut, resulting in an almost tripled lifespan of Apc(Min/+) mice (an animal model of human intestinal tumorigenesis). Mechanistically, these pseudoviruses activate the NLRP3 and AIM2 inflammasomes, leading to caspase-1-mediated tumour regression that is dependent on neither cytotoxic T lymphocytes nor humoral immune response. Blocking caspase-1 activation abrogated the therapeutic effects of the pseudoviruses. Thus, targeting innate immune sensors in tumours by the pseudoviruses might represent a strategy to treat intestinal tumours.