EMAIL THIS PAGE TO A FRIEND
Nature chemical biology
CRISPR-Cas9 strategy for activation of silent Streptomyces biosynthetic gene clusters.
PMID 28398287
Abstract
Here we report an efficient CRISPR-Cas9 knock-in strategy to activate silent biosynthetic gene clusters (BGCs) in streptomycetes. We applied this one-step strategy to activate multiple BGCs of different classes in five Streptomyces species and triggered the production of unique metabolites, including a novel pentangular type II polyketide in Streptomyces viridochromogenes. This potentially scalable strategy complements existing activation approaches and facilitates discovery efforts to uncover new compounds with interesting bioactivities.