EMAIL THIS PAGE TO A FRIEND
Cell reports
Chd8 Mutation Leads to Autistic-like Behaviors and Impaired Striatal Circuits.
PMID 28402856
Abstract
Autism spectrum disorder (ASD) is a heterogeneous disease, but genetically defined models can provide an entry point to studying the molecular underpinnings of this disorder. We generated germline mutant mice with loss-of-function mutations in Chd8, a dexa0novo mutation strongly associated with ASD, and demonstrate that these mice display hallmark ASD behaviors, macrocephaly, and craniofacial abnormalities similar to patient phenotypes. Chd8(+/-) mice display a broad, brain-region-specific dysregulation of major regulatory and cellular processes, most notably histone and chromatin modification, mRNA and protein processing, Wnt signaling, and cell-cycle regulation. We also find altered synaptic physiology in medium spiny neurons of the nucleus accumbens. Perturbation of Chd8 in adult mice recapitulates improved acquired motor learning behavior found in Chd8(+/-) animals, suggesting a role for CHD8 in adult striatal circuits. These results support a mechanism linking chromatin modification to striatal dysfunction and the molecular pathology of ASD.