EMAIL THIS PAGE TO A FRIEND
Acta cirurgica brasileira
The effects of ozone on bacterial growth and thiol-disulphide homeostasis in vascular graft infection caused by MRSA in rats.
PMID 28403346
Abstract
To investigate the microbiological, inflammatory and oxidant effects of adjuvant ozone administration in experimental rat vascular graft infection model which has not been previously investigated. Forty adult Wistar rats were divided into Sham, Control, Vancomycin, Ozone, Vancomycin+Ozone groups. Grafts were inoculated with Methicillin-resistant Staphylococcus aureus (MRSA) strain and implanted subcutaneously. Rats were treated intraperitoneally with ozone and /or intramuscularly with vancomycin for 10 days. Grafts were evaluated by quantitative bacterial cultures. Blood samples were harvested for determination of thiol-disulphide and cytokine profiles. There was no significant difference in bacterial counts between Control and Ozone Groups. In the Ozone Group median colony count was significantly higher than the Vancomycin and Vancomycin+Ozone Groups. Total thiol and disulphide levels increased and disulphide/native thiol and disulphide/total thiol ratios decreased in Ozone Group significantly. Albumin levels decreased significantly in Vancomycin and Vancomycin+Ozone Groups compared to the Sham Group. IL-1 and TNF-alpha levels significantly increased in infected rats. Decreased levels of VEGF due to infection reversed by ozone therapy in control and vancomycin groups. We didn't observe any benefit of the agent on MRSA elimination in our model. Likewise, effects of ozone on thiol-disulphide homeostasis and inflammatory cytokines were contradictory.