EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Genome editing via delivery of Cas9 ribonucleoprotein.
PMID 28410976
Abstract
The CRISPR-Cas genome editing system is very powerful. The format of the CRISPR reagents and the means of delivery are often important factors in targeting efficiency. Delivery of recombinant Cas9 protein and guide RNA (gRNA) as a preformed ribonucleoprotein (RNP) complex has recently emerged as a powerful and general approach to genome editing. Here we outline methods to produce and deliver Cas9 RNPs. A donor DNA carrying desired sequence changes can also be included to program precise sequence introduction or replacement. RNP delivery limits exposure to genome editing reagents, reduces off-target events, drives high rates of homology-dependent repair, and can be applied to embryos to rapidly generate animal models. RNP delivery thus minimizes some of the pitfalls of alternative editing modalities and is rapidly being adopted by the genome editing community.