EMAIL THIS PAGE TO A FRIEND
Photochemistry and photobiology
Multiple modes of photodynamic action by cercosporin.
PMID 2841704
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6696
Cercosporin from Cercospora hayii
C29H26O10