EMAIL THIS PAGE TO A FRIEND
World journal of microbiology & biotechnology
Bioactive compounds and antioxidant activity exhibit high intraspecific variability in Pleurotus ostreatus mushrooms and correlate well with cultivation performance parameters.
PMID 28421412
Abstract
Experimental data related with oyster mushroom production and nutritional properties usually derive from the examination of only one strain, and hence their representativeness/usefulness is questionable. This work aims at assessing intraspecific variability in Pleurotus ostreatus by studying 16 strains, under the same conditions, in respect to essential cultivation and mushroom quality aspects, and by defining the impact of intrinsic/genetic factors on such parameters. Hence, mushroom yield, earliness, crop length, biological efficiency, productivity, and their content in selected macro and microconstituents (e.g. fatty acids, sterols, individual phenolic compounds, terpenic acids, glucans) as well as their antioxidant properties (i.e., antiradical activity, ferric reducing potential, inhibition of serum oxidation) were assayed. The effect of intrinsic/genetic factors was evident, especially as regards earliness, yield of each production flush and mushroom weight, whereas biological efficiency was not particularly influenced by the cultivated strain. Moreover, phenolics, ergosterol and antiradical activity demonstrated significant variability among strains in contrast to what was observed for fatty acids, β-glucans and ferric reducing potential. The observed heterogeneity reveals the limitations of using a low number of strains for evaluating mushroom production and/or their content in bioactive compounds, and as evidenced, it is valuable for breeding and commercial purposes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774243
1,3-Propylene sulfite, 99%
C3H6O3S