EMAIL THIS PAGE TO A FRIEND
Mycologia
Six new soil-inhabiting Cladosporium species from plateaus in China.
PMID 28422608
Abstract
Cladosporium species are ubiquitous in various environments but are hitherto rarely isolated from soil. In the present study, six new Cladosporium species inhabiting the plateau soils of China are described as C. neopsychrotolerans, C. paralimoniforme, C. prolongatum, C. sinuatum, C. tianshanense, and C. verruculosum. These species are phylogenetically distinct and morphologically different from known species. This study increased the number of species classified in the C. cladosporioides and C. herbarum complexes and revealed Chinese plateau soil as a rich niche of Cladosporium species diversity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17236
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
C12H19NO3S