EMAIL THIS PAGE TO A FRIEND
Oncotarget
Parthenolide suppresses non-small cell lung cancer GLC-82 cells growth via B-Raf/MAPK/Erk pathway.
PMID 28423582
Abstract
Non-small cell lung cancer (NSCLC), one type of lung cancer, owns high rates of morbidity and mortality. B-Raf is one of the promising oncogenic drivers of NSCLC. Parthenolide, a natural product, is mainly extracted from the herbal plant Tanacetum parthenium. The effect of parthenolide on NSCLC cells and its potential as B-Raf inhibitor were studied in this study. It's shown that parthenolide exhibited the strong cytotoxicity against NSCLC cells with IC50 ranging from 6.07 ± 0.45 to 15.38 ± 1.13 μM. Parthenolide was also able to induce apoptosis, suppress proliferation and invasion in NSCLC cells. In terms of the involved mechanism, parthenolide suppressed GLC-82 cell response via targeting on B-Raf and inhibiting MAPK/Erk pathway signaling. The effect of parthenolide on B-Raf and MAPK/Erk pathway was further confirmed by RNA interference of B-Raf. Decreased expression of c-Myc in protein and mRNA level was also discovered, which is considered as the further downstream of the MAPK/Erk pathway. In addition, STAT3 activity inhibition by parthenolide contributed to its effect on GLC-82 cells, which is independent of PI3K pathway signaling and GSK3. All above provide an insight to understand the action of parthenolide as a potential B-Raf inhibitor in treatment of NSCLC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0415
Alantolactone, ≥98% (HPLC)
C15H20O2