EMAIL THIS PAGE TO A FRIEND
Journal of bioethical inquiry
Regulating the New: A Consideration of CRISPR and Approaches to Professional Standards of Practitioners of Chinese Medicine in Australia and Accessing the NDIS.
PMID 28429203