EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Membrane insertion of F0 c subunit of F0F1 ATPase depends on glycolipozyme MPIase and is stimulated by YidC.
PMID 28431927
Abstract
The F0 c subunit of F0F1 ATPase (F0-c) possesses two membrane-spanning stretches with N- and C-termini exposed to the periplasmic (extracellular) side of the cytoplasmic membrane of E. coli. Although F0-c insertion has been extensively analyzed in vitro by means of protease protection assaying, it is unclear whether such assays allow elucidation of the insertion process faithfully, since the membrane-protected fragment, an index of membrane insertion, is a full-length polypeptide of F0-c, which is the same as the protease-resistant conformation without membrane insertion. We found that the protease-resistant conformation could be discriminated from membrane-insertion by including octyl glucoside on protease digestion. By means of this system, we found that F0-c insertion depends on MPIase, a glycolipozyme involved in membrane insertion, and is stimulated by YidC. In addition, we found that acidic phospholipids PG and CL transform F0-c into a protease-resistant form, while MPIase prevents the acquisition of such a protease-resistant conformation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
861677
DL-Cysteine, technical grade
C3H7NO2S
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O