EMAIL THIS PAGE TO A FRIEND
Nature
Phase-plate cryo-EM structure of a class B GPCR-G-protein complex.
PMID 28437792
Abstract
Class B G-protein-coupled receptors are major targets for the treatment of chronic diseases, such as osteoporosis, diabetes and obesity. Here we report the structure of a full-length class B receptor, the calcitonin receptor, in complex with peptide ligand and heterotrimeric Gαsβγ protein determined by Volta phase-plate single-particle cryo-electron microscopy. The peptide agonist engages the receptor by binding to an extended hydrophobic pocket facilitated by the large outward movement of the extracellular ends of transmembrane helices 6 and 7. This conformation is accompanied by a 60° kink in helix 6 and a large outward movement of the intracellular end of this helix, opening the bundle to accommodate interactions with the α5-helix of Gαs. Also observed is an extended intracellular helix 8 that contributes to both receptor stability and functional G-protein coupling via an interaction with the Gβ subunit. This structure provides a new framework for understanding G-protein-coupled receptor function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538256
2,2-Dimethyl-1,3-propanediol, 99%
C5H12O2
C6512
Cholesteryl hemisuccinate
C31H50O4