EMAIL THIS PAGE TO A FRIEND
Oncotarget
Mutation of the Sp1 binding site in the 5' flanking region of SRY causes sex reversal in rabbits.
PMID 28445127
Abstract
Sex-determining region Y is a crucial gene that initiates male sex determination in mammals. Mutations of the Sp1-binding site in the 5' flanking region of SRY are associated with clinical male-to-female sex reversal syndrome, although such occurrences are rare and, until now, have not been reported in animal models. In this study, we mutated Sp1-binding sites in the 5' flanking region of the rabbit SRY gene using the CRISPR/Cas9 system. As expected, the SRY-Sp1 knockout rabbits had female external and internal genitalia and exhibited normal female copulatory behaviors, but they were infertile, and the adults displayed reduced follicles. Interestingly, we successfully obtained offspring from sex-reversed SRY-Sp1 knockout rabbits using embryo transfer. In summary, our study demonstrates that Sp1 is a major regulator in SRY gene transcription, and mutations of the Sp1 binding sites (Sp1-B and Sp1-C) in the 5' flanking region of SRY induce sex reversal in rabbits, which can be used as targets for clinical research of male-to-female sex reversal syndrome. Additionally, we provide the first evidence that sex reversal syndrome patients have the potential to become pregnant with the use of embryo transfer.